ContactUsLinkComponent
Masterflex® Fitting, PVDF, Straight, Hosebarb to Thread Adapters, 1/4" ID x 1/4" NPT(M); 10/PK
Masterflex® Fitting, PVDF, Straight, Hosebarb to Thread Adapters, 1/4" ID x 1/4" NPT(M); 10/PK
Catalog # MFLX30615-16
CE certificate
undefined
Masterflex® Fitting, PVDF, Straight, Hosebarb to Thread Adapters, 1/4" ID x 1/4" NPT(M); 10/PK
Catalog # MFLX30615-16
CE certificate
About this item
Quick and easy installation with no tools required.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 1/4" ID x 1/4" NPT(M); 10/PK
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). PVDF material features excellent chemical resistance with a wide temperature range. Sterilize via ethylene oxide or autoclave.
  • Barb size/tubing int.Ø (port 1):
    0,25"
  • Connection Type:
    Barb x NPT(M)
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 2):
    0,25"
  • Description:
    Kynar BARB,NPT,L/S 17/24 & I/P26,10/
  • Seq:
    03
  • Environmentally Preferable:
  • Model:
    30615-16
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)
  • Pk:
    10
  • Fitting material:
    PVDF
Frequently Bought Together