ContactUsLinkComponent
Masterflex® Antisiphon Kit for Syringe Pump, Infusion and Withdrawal Design
Masterflex® Antisiphon Kit for Syringe Pump, Infusion and Withdrawal Design
Catalog # MFLX74905-72
undefined
Masterflex® Antisiphon Kit for Syringe Pump, Infusion and Withdrawal Design
Catalog # MFLX74905-72
  • Model:
  • 74905-62
  • 74905-61
  • 74905-60
  • Syringe size accepted:
  • 0,5 µl - 10 ml
  • 60 - 140 ml
  • 0,5 µl - 60 ml
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • Currently Viewing
Specifications
About this item
Make the most of your Masterflex® syringe pumps with these accessories.
  • Optimize performance of your Masterflex pumps
Parts and accessories for Masterflex syringe pumps.
  • Description:
    Antisiphon Kit for Syringe Pump, Infusion and Withdrawal design
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together