SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
RECOMBINANT RAT TGF-BETA RII/TGFBR2 PROTEIN (HFC TAG) 100UG 1 * 100 µG
SINO80139-R02H-100
:
Sino Biological
RECOMBINANT RAT TGF-BETA RII/TGFBR2 PROTEIN (HFC TAG) 100UG 1 * 100 µG
SINO80139-R02H-100
:
Sino Biological
Frequently Bought Together
Transfer pipettes
: RATIOLAB
Sealing film, Axygen® AxySeal
: Corning
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Multiwell Cell Culture Plates
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Aluminium Foil Seals for Cold Storage
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF