Recombinant Human IL17A & IL17F Heterodimer Protein 1 * 20 µG
Catalog # SINOCT047-HNAE-20
Supplier: Sino Biological
Recombinant Human IL17A & IL17F Heterodimer Protein 1 * 20 µG
Catalog # SINOCT047-HNAE-20
Supplier: Sino Biological
Frequently Bought Together
Cryogenic vials, Cryovial®
Supplier: Simport Scientific
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Dewar flasks, Nalgene®
Supplier: Thermo Fisher Scientific
Containers with optional snap caps, Nunc™
Supplier: Thermo Fisher Scientific
Blotting transfer papers
Supplier: Ahlstrom-Munksjö
PCR Thermo-Tube and cap strips, 0.2 ml, ABgene®
Supplier: Thermo Fisher Scientific
VWR®, Deep Well Plates, 96 Square Well, Reinforced for Genomics Applications
Supplier: VWR Collection
Microcentrifuge tubes, 1,5 ml, in resealable bags, SuperSpin®
Supplier: LABCON
122263, triple-packed 1 * 200 items
Supplier: Greiner Bio-One
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Inoculating loops, stainless steel
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Storage cryobox, Nalgene®
Supplier: Thermo Fisher Scientific
Storage Boxes
Supplier: EPPENDORF
Serological pipettes
Supplier: Greiner Bio-One
Crystallisation plates, CrystalStar™, 96-well CrystalQuick™ and CrystalDrop™
Supplier: Greiner Bio-One
VWR® Square Media Biotainer Canisters
Supplier: VWR Collection