Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant Human GAD65/GAD2 Protein (GST Tag) 1 * 100 µG
Catalog # SINO17802-H09B-100
Supplier:
Sino Biological
Expand image
Recombinant Human GAD65/GAD2 Protein (GST Tag) 1 * 100 µG
Catalog # SINO17802-H09B-100
Supplier:
Sino Biological
Frequently Bought Together
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF