Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Fall Arrest Roofing Kits

Fall Arrest Roofing Kits

Nylon drawstring bag
Catalog # PRTWFP99BKR
Supplier:   Portwest
Fall Arrest Roofing Kits
Catalog # PRTWFP99BKR
Supplier:  Portwest
Fall Arrest Roofing Kits
Fall Arrest Roofing Kits

Nylon drawstring bag

Price
SFr. 15.40
Log in for availability
1 * 1 ST
Product Details & Documents

Kit includes two-point harness, carabiner, 12 mm detachable rope grab, 10 m kernmantle static rope, and nylon drawstring bag.

  • Chest loops
  • High-quality 12 mm kernmantle rope
  • Maximum rated load: 100 kg

The versatile drawstring bag is high-strength yet lightweight, making it ideal for securely holding fall protection products.

10-year shelf life from manufacturing date outlined on product label.

Specifications
  • Description:
    Nylon drawstring bag
  • Seq:
    02