3-AMINO-3-METHYL-1-PHENYLBUTAN-2-ONE HYDROCHLORIDE[HCL], 95% 1 * 1 g
Catalog # UKROBBV-45399133
Anbieter: UKRORGSYNTHESIS
3-AMINO-3-METHYL-1-PHENYLBUTAN-2-ONE HYDROCHLORIDE[HCL], 95% 1 * 1 g
Catalog # UKROBBV-45399133
Anbieter: UKRORGSYNTHESIS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Anbieter: VWR Chemicals
Ethanol, GPR RECTAPUR®
Anbieter: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Anbieter: VWR Chemicals