ContactUsLinkComponent
Masterflex® Dispensing Gun PTFE 1/2"npt (Spray Gun Valves (actuated))
Masterflex® Dispensing Gun PTFE 1/2"npt (Spray Gun Valves (actuated))
Catalog # MFLX98515-07
undefined
Masterflex® Dispensing Gun PTFE 1/2"npt (Spray Gun Valves (actuated))
Catalog # MFLX98515-07
About this item
No splashing.
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.
  • Description:
    Dispensing Gun PTFE 1/2"npt (spray gun valves (actuated))
  • Environmentally Preferable:
  • Wetted materials:
    PTFE
  • Type:
    98515-07
  • Warranty:
    1 year
  • Connection type:
    NPT(F)
  • Pk:
    1
Frequently Bought Together