ContactUsLinkComponent
Masterflex® Spray Gun, PTFE for DI Water, 3/8" NPT(F); 1/Ea
Masterflex® Spray Gun, PTFE for DI Water, 3/8" NPT(F); 1/Ea
Catalog # MFLX98515-01
undefined
Masterflex® Spray Gun, PTFE for DI Water, 3/8" NPT(F); 1/Ea
Catalog # MFLX98515-01
About this item
No splashing.
  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.
  • Description:
    Spray gun, PTFE for DI water; 3/8"
  • Environmentally Preferable:
  • Wetted materials:
    PTFE
  • Type:
    98515-01
  • Warranty:
    1 year
  • Connection type:
    NPT(F)
  • Pk:
    1
Frequently Bought Together