Universal Drum Wrench, Dual-Ended, Aluminum
Codice catalogo MFLX70607-92
Nome del fornitore: Avantor Fluid Handling
Specifiche
Vedi tutte le configurazioni del prodotto- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Attualmente in visualizzazione
Specifiche
Informazioni su questo articolo
Remove or tighten plugs on most industrial drums.
- Fiberglass-reinforced nylon wrenches are lightweight and easy to carry
- Use the nonsparking brass alloy models for opening drums containing flammables
- More corrosion resistant than steel versions
- 70607-92 is made of die cast aluminium
Use the round end for outside splines of bung caps. Use the long end for internal splines of bung caps.
Round end for outside splines of bung caps. Long end for internal splines of bung caps.
- Description:Universal Drum Wrench, Dual-Ended, Aluminum
- Environmentally Preferable:
- Warranty:1 year
- Pk:1
Frequently Bought Together