Auftragserfassung
Switzerland
ContactUsLinkComponent
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.175" Diameter
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.175" Diameter
Catalog # MFLX17315-24
undefined
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.175" Diameter
Catalog # MFLX17315-24

About this item

Mix, react, and/or separate liquid-liquid mixtures in a single device.

  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations

Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.

  • Ø:
    2,9845 cm
  • Bezeichnung:
    Centrifugal Multi-Purpose Separator/Extractor Weir; 1,175" Diameter
  • Environmentally Preferable:
  • Garantie:
    2 years
  • VE:
    1
Frequently Bought Together