CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.075" Diameter
Catalog # MFLX17315-20
Supplier: Avantor Fluid Handling
About this item
Mix, react, and/or separate liquid-liquid mixtures in a single device.
- Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
- Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.
- Ø:2,7305 cm
- Description:Centrifugal Multi-Purpose Separator/Extractor Weir; 1,075" Diameter
- Environmentally Preferable:
- Warranty:2 years
- Pk:1
Frequently Bought Together