ContactUsLinkComponent
Drum Pump, 8, 10 Or 12 Oz Per Stroke, 316SS Rod
Drum Pump, 8, 10 Or 12 Oz Per Stroke, 316SS Rod
Catalog # MFLX06511-60
undefined
Drum Pump, 8, 10 Or 12 Oz Per Stroke, 316SS Rod
Catalog # MFLX06511-60
About this item
Economical pumps for use in cabinets or tight work spaces.
  • Fit 15, 30, and 55-gallon drums with 2" IPS bung openings
  • Designed for use in industrial, agricultural, labs, and automotive industries
Choose from center-level pump and lever-action pump model options.
Center-Lever Pump is self-priming and features an anti-drip spout to eliminate wasted fluid. This pump is ideal for oils, detergents, and other compatible industrial fluids.
Lever-Action Pumps feature a curved discharge spout for convenient hose extension. Choose from three models depending on your chemical needs. Model 06511-50 is ideal for water, alcohol, gasoline, diesel, kerosene, and lubricants. Model 06511-60 handles the same fluids, plus water-based chemicals, acids, and alkalis. Model 06511-80 handles the same fluids, plus strong acids and strong alkalis.
  • Flow rate:
    16 strokes/gal.
  • Description:
    Drum Pump, 8, 10 Or 12 Oz Per Stroke, 316SS Rod
  • Environmentally Preferable:
  • Wetted materials:
    Ryton® body, Stainless Steel rod with Viton seals
  • Tube length, inlet:
    36,5"
  • Warranty:
    3 Months
  • Pk:
    1
Frequently Bought Together