ContactUsLinkComponent
Micropump 81980 GEAR PUMP HEAD0.316 ML/REV
Micropump 81980 GEAR PUMP HEAD0.316 ML/REV
Catalog # MFLX74012-11
Micropump 81980 GEAR PUMP HEAD0.316 ML/REV
Catalog # MFLX74012-11
About this item
Offering remote C-mount choices for your low-flow applications.
  • Use for applications requiring high initial differential pressure.
  • Suction-shoe design ensures complete fluid transfer for increased volumetric accuracy
  • Pump efficiency increases as discharge pressure increases
The compact pump head requires a drive for proper operation. Order drives separately.
Optional Service kit includes: gears, suction shoe, shaft seals, and springs. Order separately.
Contact our Application Specialists for assistance.
  • Description:
    Low-flow pump head, pressure-loaded, 0,316 ml/revolution
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together