ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Threaded Adapter, 1/4-28 UNF; 25/PK
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Threaded Adapter, 1/4-28 UNF; 25/PK
Catalog # MFLX45518-82
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Threaded Adapter, 1/4-28 UNF; 25/PK
Catalog # MFLX45518-82
CE certificate
About this item
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Connection Type: Male Luer Lock to Threaded, 1/4-28 UNF
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
  • Connection Type:
    Male Luer to Thread
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Male Luer Lock to Threaded Adapter, ¼-28 UNF; 25/PK
  • Seq:
    07
  • Environmentally Preferable:
  • Model:
    45518-82
  • Connection type (port 1):
    Male luer lock
  • Connection type (port 2):
    ¼-28 UNF
  • No. of ports:
    2
  • Pk:
    25
  • Fitting material:
    Polypropylene
  • Thread size UNF(M) (port 1):
    ¼"-28
Frequently Bought Together