Auftragserfassung
Switzerland
ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PK
  MFLX45508-60
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, 10-32 UNF; 25/PK
  MFLX45508-60
CE certificate
Leichter Übergang zwischen Luer-Fittings und UNF- oder NPT-Gewinde.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Diese Adapter biete einen Übergang zwischen Luer-Fittings und UNF- oder NPT-Gewinde. Außen-Luer- wie Innen-Luer-Verbindungen sind verfügbar. Alle Gewindeverbindungen sind Innengewinde.
  • Verbindungstyp:
    Female Luer to Thread
  • Flow pattern:
    Straight
  • Bezeichnung:
    Innen-Luer x 10-32 UNF-Gewinde, PP, 25/Pck 45508-60
  • Seq:
    04
  • Environmentally Preferable:
  • Modell:
    45508-60
  • Connection type (port 1):
    Female luer
  • Connection type (port 2):
    10-32 UNF thread
  • No. of ports:
    2
  • VE:
    25
  • Fitting material:
    Polypropylene
  • Thread size UNF(M) (port 1):
    10-32
Frequently Bought Together