ContactUsLinkComponent
Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/Pk
Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/Pk
Catalog # MFLX45501-66
CE certificate
Avantor exclusive
undefined
Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/Pk
Catalog # MFLX45501-66
CE certificate
Avantor exclusive
About this item
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
  • Connection Type:
    Female Luer to Thread
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/Pk
  • Seq:
    03
  • Environmentally Preferable:
  • Model:
    45501-66
  • Connection type (port 1):
    Female luer
  • No. of ports:
    2
  • Warranty:
    3 Months
  • Pk:
    25
  • Fitting material:
    Polycarbonate
  • Thread size UNF(M) (port 1):
    1/4"-28
Frequently Bought Together