ContactUsLinkComponent
Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 1/8" ID x 10-32 UNF(M); 100/PK
Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 1/8" ID x 10-32 UNF(M); 100/PK
Catalog # MFLX40625-05
CE certificate
undefined
Masterflex® Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 1/8" ID x 10-32 UNF(M); 100/PK
Catalog # MFLX40625-05
CE certificate
About this item
No tools needed for assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
  • Barb size/tubing int.Ø (port 2):
    1/8"
  • Connection Type:
    Thread to Barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, HDPE, Straight, Hosebarb to Thread Adapter, 1/8" ID x 10-32 UNF(M); 100/PK
  • Seq:
    17
  • Environmentally Preferable:
  • Model:
    40625-05
  • Sterilisation:
    Ethylene Oxide (ETO)
  • Pk:
    100
  • Fitting material:
    HDPE
  • Thread size UNF(M) (port 2):
    10-32"
Frequently Bought Together