ContactUsLinkComponent
Masterflex® Ultrasonic Flow Sensor Communications Package
Masterflex® Ultrasonic Flow Sensor Communications Package
Catalog # MFLX33501-96
undefined
Masterflex® Ultrasonic Flow Sensor Communications Package
Catalog # MFLX33501-96
About this item
Make the most of your Masterflex flowmeters with these replacement parts and other accessories.
  • Optimize performance of your flowmeter with these accessories
These power supplies, adapters, and other accessories are suitable for use with your Masterflex flowmeter and flow sensors.
  • Description:
    Masterflex Ultrasonic flow sensor communications package
  • Seq:
    07
  • Environmentally Preferable:
  • Model:
    33501-96
  • Connections:
    One USB connects to computor for power and data, 2nd USB for addtional power, M12 attaches to Masterflex Flow Sensor
  • Warranty:
    1 year
  • Compatible with:
    33501-13, 33501-14, 33501-15, 33501-16, 33501-17, 33501-18, 33501-24, 33501-25, 33501-26, 33501-35, 33501-36, 33501-73
  • Pk:
    1
Frequently Bought Together