ContactUsLinkComponent
Masterflex® Fitting, HDPE, Y-Connector, Hosebarb Reducer, 3/16" ID x 3/16" ID x 5/16" ID; 10/PK
Masterflex® Fitting, HDPE, Y-Connector, Hosebarb Reducer, 3/16" ID x 3/16" ID x 5/16" ID; 10/PK
Catalog # MFLX30726-12
undefined
Masterflex® Fitting, HDPE, Y-Connector, Hosebarb Reducer, 3/16" ID x 3/16" ID x 5/16" ID; 10/PK
Catalog # MFLX30726-12
  • Model:
  • 30726-12
  • 30726-11
  • 30726-37
  • 30726-35
  • 30726-36
  • 30726-17
  • 30726-14
  • 30726-16
  • 30726-13
  • 30726-52
  • 30726-53
  • 30726-51
  • 30726-56
  • 30726-57
  • 30726-54
  • 30726-55
  • 30726-34
  • 30726-31
  • 30726-32
  • 30726-33
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Ethylene oxide (ETO) or gamma
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Currently Viewing
Specifications
About this item
Ensure a tight seal every time.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
  • Barb size/tubing int.Ø (port 1):
    3/16"
  • Barb size/tubing int.Ø (port 2):
    3/16"
  • Connection Type:
    Barbed Reducer
  • Flow pattern:
    Y
  • Description:
    Fitting, HDPE, Y-Connector, Hosebarb Reducer, 3/16" ID x 3/16" ID x 5/16" ID; 10/PK
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    30726-12
  • Sterilisation:
    Ethylene oxide (ETO) only
  • Pk:
    10
  • Barb size/tubing int.Ø (port 3):
    5/16"
  • Fitting material:
    HDPE
Frequently Bought Together