ContactUsLinkComponent
Masterflex® Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 3/8" ID; 1/EA
Masterflex® Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 3/8" ID; 1/EA
Catalog # MFLX30620-24
undefined
Masterflex® Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 3/8" ID; 1/EA
Catalog # MFLX30620-24
  • Model:
  • 30620-24
  • 31209-57
  • 30620-22
  • 31209-56
  • 30620-23
  • 30620-21
  • 31209-55
  • Orifice dimensions:
  • 0,28125"
  • 0,03125"
  • 0,0625"
  • Sterilisation:
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
  • 1 Month
  • 1 year
  • 1 Month
  • 1 year
  • 1 Month
  • 1 Month
  • 1 year
  • Currently Viewing
Specifications
About this item
Ensure a tight seal every time.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Designed for quick and easy assembly with flexible tubing
  • Connection Type: 3/8" ID; 1/EA
Combine these simple, economical fittings with flexible tubing for use in your low pressure applications.
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing.
  • Barb size/tubing int.Ø (port 1):
    3/8"
  • Barb size/tubing int.Ø (port 2):
    3/8"
  • Connection Type:
    Barbed Union
  • Flow pattern:
    Y
  • Description:
    Fitting, Stainless Steel, Y-Connector, Hose Barb Union, 3/8" ID; 1/EA
  • Seq:
    04
  • Environmentally Preferable:
  • Model:
    30620-24
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
    1 Month
  • Pk:
    1
  • Barb size/tubing int.Ø (port 3):
    3/8"
  • Fitting material:
    Stainless Steel 316
Frequently Bought Together