ContactUsLinkComponent
Masterflex® Fitting, Polycarbonate, Three-Way Stopcock, Male Luer Lock, Non-Sterile; 10/PK
Masterflex® Fitting, Polycarbonate, Three-Way Stopcock, Male Luer Lock, Non-Sterile; 10/PK
Catalog # MFLX30600-02
undefined
Masterflex® Fitting, Polycarbonate, Three-Way Stopcock, Male Luer Lock, Non-Sterile; 10/PK
Catalog # MFLX30600-02
About this item
Regulate flow without any leaking.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Three-way stopcock/flow diverter with male luer lock connections
  • HDPE handle rotates 180°
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
  • Flow pattern:
    Three-Way
  • Description:
    Fitting, Polycarbonate, Three-Way Stopcock, Male Luer Lock, Non-Sterile; 10/PK
  • Environmentally Preferable:
  • Wetted materials:
    Polycarbonate
  • Warranty:
    Out of box
  • Pk:
    10
  • Inlet port:
    Female Luer
  • Outlet port:
    Male Luer Lock
Frequently Bought Together