ContactUsLinkComponent
Masterflex® Ismatec® Cartridge Holder
Masterflex® Ismatec® Cartridge Holder
Catalog # MFLX07618-73
Masterflex® Ismatec® Cartridge Holder
Catalog # MFLX07618-73
About this item
Choose new cassettes based upon material of construction, tubing type and pressure capabilities.
  • Material of construction standard POM-C or optional PVDF
There are two different designs of cartridge. The Click-N-Go design allows for tubing to be used in systems that have up to 1 bar of back pressure exerted on the tubing. The Pressure Lever design allows for system pressures up to 1.5 bar.
  • Description:
    Cartridge holder
  • Seq:
    01
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    2
Pressure lever cassettes with inserts
Product Information
Frequently Bought Together