ContactUsLinkComponent
Masterflex® L/S® Easy-Load® Pump Head Replacement Open-Head Sensor for 07514-series
Masterflex® L/S® Easy-Load® Pump Head Replacement Open-Head Sensor for 07514-series
Catalog # MFLX07514-08
1 year warranty
undefined
Masterflex® L/S® Easy-Load® Pump Head Replacement Open-Head Sensor for 07514-series
Catalog # MFLX07514-08
1 year warranty
  • Model:
  • 07514-08
  • 07019-80
  • 07019-79
  • 07014-81
  • 07017-91
  • 07019-83
  • 07017-81
  • 07019-82
  • 07019-75
  • 07035-91
  • 07019-74
  • 07518-69
  • 77602-18
  • 07013-95
  • 07019-78
  • 07036-02
  • 07016-91
  • 07016-81
  • 07019-77
  • 07036-01
  • 07019-73
  • 07014-91
  • 07013-81
  • 07013-91
  • 07019-85
  • 07013-76
  • 07514-18
  • 07019-92
  • 07015-81
  • 07015-80
  • 07015-91
  • 07516-08
  • 07015-90
  • 07516-07
  • 07018-91
  • 07018-81
  • 07015-75
  • 07013-80
  • 07516-18
  • 07516-17
  • 77602-08
  • 07518-68
  • 07024-91
  • 07024-81
  • No. of channels:
  • 2
  • 1
  • Open-head sensor:
  • Yes
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • No
  • Pump series:
  • L/S
  • I/P
  • I/P
  • L/S
  • L/S
  • I/P
  • L/S
  • I/P
  • I/P
  • L/S
  • I/P
  • L/S
  • I/P
  • L/S
  • I/P
  • L/S
  • L/S
  • L/S
  • I/P
  • L/S
  • I/P
  • L/S
  • L/S
  • L/S
  • I/P
  • L/S
  • I/P
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • I/P
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Tubing sizes accepted:
  • I/P 73
  • L/S 14
  • L/S 17
  • I/P 73, 82
  • L/S 17
  • I/P 73, 82
  • I/P 82
  • L/S 35
  • I/P 82
  • L/S 13, 14, 16, 25, 17, 18
  • I/P 73
  • L/S 36
  • L/S 16
  • L/S 16
  • I/P 82
  • L/S 36
  • I/P 73
  • L/S 14
  • L/S 13
  • L/S 13
  • I/P 73, 82
  • L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
  • I/P
  • L/S 15
  • L/S 15, 24, 35, 36
  • L/S 15
  • L/S 15, 24, 35, 36
  • L/S 18
  • L/S 18
  • L/S 15, 24
  • L/S 13, 14, 16, 25, 17, 18
  • L/S® 15, L/S 24
  • L/S® 15, L/S 24
  • L/S 24
  • L/S 24
  • Warranty:
  • 1 year
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 3 Months
  • 2 years
  • 2 years
  • 1 year
  • 2 years
  • 2 years
  • 2 years
  • 1 year
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 1 year
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 3 Months
  • 1 year
  • 1 year
  • 2 years
  • 2 years
  • Currently Viewing
Specifications
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • Pump series:
    L/S
  • Description:
    L/S® Easy-Load® Pump Head Replacement Open-Head Sensor for 07514-series
  • Seq:
    35
  • Environmentally Preferable:
  • Model:
    07514-08
  • Open-head sensor:
    Yes
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together