ContactUsLinkComponent
Masterflex® L/S® Standard Pump Head, Mounting Hardware, Mounts One L/S 36 Pump Head, Stainless Steel; 2/Set
Masterflex® L/S® Standard Pump Head, Mounting Hardware, Mounts One L/S 36 Pump Head, Stainless Steel; 2/Set
Catalog # MFLX07036-01
undefined
Masterflex® L/S® Standard Pump Head, Mounting Hardware, Mounts One L/S 36 Pump Head, Stainless Steel; 2/Set
Catalog # MFLX07036-01
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
  • Suitable for use with L/S® Standard Pump Heads 07036-30 and -31
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • No. of channels:
    1
  • Pump series:
    L/S
  • Description:
    Standard Pump Head, Mounting Hardware, Mounts One L/S® 36 Pump Head, Stainless Steel; 2/Set
  • Seq:
    33
  • Environmentally Preferable:
  • Model:
    07036-01
  • Open-head sensor:
    No
  • Tubing sizes accepted:
    L/S 36
  • Warranty:
    1 year
  • Pk:
    1
Frequently Bought Together