ContactUsLinkComponent
Masterflex® I/P® Standard Pump Head, Replacement Rotor Assembly, Long Shaft, SS Rotor
Masterflex® I/P® Standard Pump Head, Replacement Rotor Assembly, Long Shaft, SS Rotor
Catalog # MFLX07019-83
undefined
Masterflex® I/P® Standard Pump Head, Replacement Rotor Assembly, Long Shaft, SS Rotor
Catalog # MFLX07019-83
About this item
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
  • Suitable for use with I/P® Standard pump heads 07019-01, -41, -26, -51.
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • Pump series:
    I/P
  • Description:
    Masterflex I/P® Standard Pump Head, Replacement Rotor Assembly, Long Shaft, SS Rotor; 1/Ea
  • Seq:
    27
  • Environmentally Preferable:
  • Model:
    07019-83
  • Open-head sensor:
    No
  • Tubing sizes accepted:
    I/P 73, 82
  • Warranty:
    2 years
  • Pk:
    1
  • Rotor material:
    Stainless Steel
Frequently Bought Together