ContactUsLinkComponent
Masterflex® I/P® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, SS Rotor, I/P® 82
  MFLX07019-75
undefined
Masterflex® I/P® Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, SS Rotor, I/P® 82
  MFLX07019-75
Get the most our of your Masterflex® Pump Head with these replacement parts.
  • Choose replacement parts suitable for your L/S® and I/P® pump head
  • Suitable for use with I/P® 82 Standard pump head
  • Constructed with durable polycarbonate (PC) housing and stainless steel (SS) rotor
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
  • Pump series:
    I/P
  • Description:
    Standard Pump Head, Replacement End Bell Assembly, Polycarbonate Housing, SS Rotor, I/P® 82 Tubing
  • Seq:
    21
  • Environmentally Preferable:
  • Model:
    07019-75
  • Open-head sensor:
    No
  • Tubing sizes accepted:
    I/P 82
  • Warranty:
    2 years
  • Pk:
    1
  • Housing material:
    PC
  • Rotor material:
    Stainless Steel
Frequently Bought Together