ContactUsLinkComponent
Masterflex® Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/Pk
Masterflex® Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/Pk
Catalog # MFLX06833-01
undefined
Masterflex® Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/Pk
Catalog # MFLX06833-01
About this item
Won't rust, corrode, or distort.
  • Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
  • Housing material:
    Polyoxymethylene (POM) Acetal
  • Description:
    Mini Tubing Clamps for up to 1/4" Tubing, On/Off, Acetal; 25/Pk
  • Environmentally Preferable:
  • Pinch valve tube size:
    1/16" to 1/4" OD
  • Pk:
    25
Frequently Bought Together