À propos de cet article
Safely transfer, sample, and store biopharmaceutical products and reagents.
- Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to lose connections
- Assemblies are pre-sterilized by gamma irradiation—eliminates labor and validation costs associated with assembly and cleaning
Cap assembly includes a seamless liner with premolded C-Flex® fill and vent tubes to create an unobstructed fluid path. The 18" fill tube is plugged at the terminal end to maintain sterility until use. Vent tube includes a 0.2-micron PTFE vent filter to prevent infiltration by contaminants. Vent filter is hydrophobic to guard against loss of costly product or media during filling or emptying. The one-piece liner eliminates the risk of bacterial entrapment and contamination and forms an elastomeric seal between cap and bottle rim.
C-Flex tubing is manufactured according to cGMP and meets USP Class VI and USDA requirements. Tubing can withstand temperatures from –100 to 275 °F (–73 to 135 °C). C-Flex tubing has exceptional tensile and tear strength, extremely low extractables, and excellent biocompatibility. The smooth inner surface reduces absorption and protein binding.
- Cap Size:53B
- Capacity:250 ml
- Description:Gamma Irradiated Sampling Bottle Assembly, Sterile, PETG Bottle; 250mL
- Material:Polyethylene terephtalate (PET)
- Environmentally Preferable:
- Special feature:Sterile
- Int. Ø tubing:1/8" ID x 1/4" OD
- Warranty:3 Months
- Cap colour:Natural
- Pk:1
- Bottle colour:Clear