BUFFER FB FOR NUCELOSPIN FINISHER MIDI K 1 * 1.000 mL
Catalog # MANA740438.1000
Supplier: MACHEREY-NAGEL
BUFFER FB FOR NUCELOSPIN FINISHER MIDI K 1 * 1.000 mL
Catalog # MANA740438.1000
Supplier: MACHEREY-NAGEL
Frequently Bought Together
PIROCTONE OLAMINE 1 * 500 mg
Supplier: Merck
Methacrylamide
Supplier: EHRENSTORFER
Imiquimod Related Compounds, Reference Standard
Supplier: USP
Michigan Pesticide Mixture 2
Supplier: EHRENSTORFER
Methyl Palmitate, Reference Standard
Supplier: USP
Heptachlor-exo-epoxide (cis-, isomer B) 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Diphenhydramine Hydrochloride, Reference Standard
Supplier: USP
N-Nitroso-ethyl methylamine 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
Disulphoton sulphoxide Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
CHLOROFORM-D 1 * 10 g
Supplier: LGC Standards PROMOCHEM
Halfenprox 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
Accessories for ion selective meters, LAQUAtwin
Supplier: HORIBA
Supelco® PESTANAL® Organic Reference Standard, Oxamyl
Supplier: Merck
OEKO-TEX PAH Mixture
Supplier: EHRENSTORFER
pH buffer solutions, colour coded
Supplier: Hach