Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Rubbermaid® Step Stools

Rubbermaid® Step Stools

Black, 406×343×27 mm
Catalog # 129-1871
Rubbermaid® Step Stools
Catalog # 129-1871
Rubbermaid® Step Stools
Rubbermaid® Step Stools

Black, 406×343×27 mm

Price
156.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Heavy-duty stepstool glides easily on retractable rubber casters and supports loads up to 158,8 kg.

  • Soft, slip-resistant pads on both steps
  • Wheels retract when stepped on
  • Material: Plastic

Heavy-duty plastic construction will not damage walls or furniture.

Specifications
  • Colour:
    Black
  • Seq:
    02
  • W×D×H:
    406×343×27 mm