Order Entry
Switzerland
Orders LinkContactUsLinkComponent
pH/conductivity meter, SevenDirect SD23

pH/conductivity meter, SevenDirect SD23

pH/Conductivity meter SevenDirect SD23 Pure H₂O
Catalog # 662-2412
Supplier:   Mettler - Toledo
pH/conductivity meter, SevenDirect SD23
Catalog # 662-2412
Supplier:  Mettler - Toledo
pH/conductivity meter, SevenDirect SD23
pH/conductivity meter, SevenDirect SD23

pH/Conductivity meter SevenDirect SD23 Pure H₂O

Price
SFr. 4’275.00
Log in for availability
1 * 1 ST
Product Details & Documents

Robust bench top meter for accurate pH and conductivity determinations in the lab and near production.

  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement, also prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)

The large colour touch screen offers direct menu access, fast data entry, and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample also sensor and method IDs with every measurement.

Specifications
  • Description:
    pH/Conductivity meter SevenDirect SD23 Pure H₂O
  • Seq:
    03
  • mV range:
    −2000,0 - +2000,0
  • pH range:
    −2,000 - +20,000
  • Conductivity resolution:
    Auto-range, max. 0,001
  • Calibration:
    pH: Linear, Segmented
    Conductivity: Linear, Manual cell constant
  • mV resolution:
    1 / 0,1
  • Temperature resolution:
    0,1 °C
  • Temperature accuracy:
    ±0,1 (0...100 °C)
  • Resistivity range:
    0,00 - 100,0 MΩ*cm
  • Cat. No.:
    662-2412
  • Conductivity accuracy:
    0,5% of measured value
  • pH accuracy:
    ±0,002
  • pH resolution:
    0,1 / 0,01 / 0,001
  • Conductivity range:
    0,000 μS/cm - 2000 mS/cm
  • TDS range:
    0,00 mg/L - 1000 g/L
  • Temperature range:
    −30,0…+130,0 °C
  • mV accuracy:
    ±0,1 (−500,0 - +500,0 mV) / ±0,2 (−2000,0 - +2000,0 mV)
  • Calibration points:
    pH: 1 - 5;
    Conductivity: 1 or 2
  • Salinity range:
    0,00 - 80,00 psu
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Replaceable protective cover for SevenDirect meters
    Replaceable protective cover for SevenDirect meters
    662-2382
    Log in for availability
    1 * 1 ST - 58.30 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Screws and feet set EasyPlace
    Screws and feet set EasyPlace
    662-2383
    Log in for availability
    1 * 1 ST - 31.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Electrode arm, EasyPlace Complete supports efficient measurement and calibration; With perfect vertical movement, the risk of any spillage is minimised; Includes parking beaker, parking beaker insert and sachet holder
    Electrode arm, EasyPlace Complete supports efficient measurement and calibration; With perfect vertical movement, the risk of any spillage is minimised; Includes parking beaker, parking beaker insert and sachet holder
    662-2413
    Log in for availability
    1 * 1 ST - 352.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Magnetic stirrer, EasyMix for use with SevenDirect meters; The EasyMix can be controlled manually or automatically by the SevenDirect meter
    Magnetic stirrer, EasyMix for use with SevenDirect meters; The EasyMix can be controlled manually or automatically by the SevenDirect meter
    662-2414
    Log in for availability
    1 * 1 ST - 417.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Parking beaker, EasyPlace electrode arm; Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements; or use it to store the wetting cap while measuring
    Parking beaker, EasyPlace electrode arm; Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements; or use it to store the wetting cap while measuring
    662-2415
    Log in for availability
    1 * 1 ST - 26.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Sachet holder, EasyPlace rotating; hold up to three buffer sachets and prevent spillage; It can be rotated to place one buffer after the other in the calibration position
    Sachet holder, EasyPlace rotating; hold up to three buffer sachets and prevent spillage; It can be rotated to place one buffer after the other in the calibration position
    662-2417
    Log in for availability
    1 * 1 ST - 50.30 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Parking beaker insert, EasyPlace; should be used while using the parking beaker with storage solution
    Parking beaker insert, EasyPlace; should be used while using the parking beaker with storage solution
    662-2416
    Log in for availability
    1 * 1 ST - 9.80 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Sensor holder, EasyPlace Single for the EasyPlace electrode arm holds one sensor
    Sensor holder, EasyPlace Single for the EasyPlace electrode arm holds one sensor
    662-2418
    Log in for availability
    1 * 1 ST - 26.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Sensor Holder, EasyPlace Triple for the EasyPlace electrode arm holds up to three sensors
    Sensor Holder, EasyPlace Triple for the EasyPlace electrode arm holds up to three sensors
    662-2419
    Log in for availability
    1 * 1 ST - 31.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Sensor Holder, EasyPlace Single Extended for the EasyPlace electrode arm with extended length for use with large sample vessels. Holds one sensor
    Sensor Holder, EasyPlace Single Extended for the EasyPlace electrode arm with extended length for use with large sample vessels. Holds one sensor
    662-2420
    Log in for availability
    1 * 1 ST - 31.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Pole EasyPlace, 30 cm, standard pole for the EasyPlace electrode arm
    Pole EasyPlace, 30 cm, standard pole for the EasyPlace electrode arm
    662-2421
    Log in for availability
    1 * 1 ST - 251.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Pole, EasyPlace 50 cm, extended pole for the EasyPlace electrode arm for use with large sample vessels
    Pole, EasyPlace 50 cm, extended pole for the EasyPlace electrode arm for use with large sample vessels
    662-2422
    Log in for availability
    1 * 1 ST - 302.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Base plate for the EasyPlace electrode arm
    Base plate for the EasyPlace electrode arm
    662-2423
    Log in for availability
    1 * 1 ST - 151.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Power supply, 18 W, does not include country-specific plug (order plugs separately with model (662-2426)
    Power supply, 18 W, does not include country-specific plug (order plugs separately with model (662-2426)
    662-2424
    Log in for availability
    1 * 1 ST - 100.50 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Top cover pole, EasyPlace or the top of the EasyPlace pole, has two positions for long-term sensor storage
    Top cover pole, EasyPlace or the top of the EasyPlace pole, has two positions for long-term sensor storage
    662-2425
    Log in for availability
    1 * 1 ST - 26.00 SFr.

Product Information

  • Catalog #
    Availability
    Price
  • Plug set, set of country-specific plugs for Power supply 18 W (662-2424)
    Plug set, set of country-specific plugs for Power supply 18 W (662-2424)
    662-2426
    Log in for availability
    1 * 1 ST - 34.10 SFr.