ContactUsLinkComponent
XAN TEC SPR Sensorchip. Biacore Series S kompatibel. Beschichtung: 200 nm Biotin derivatisiertes Carboxymethyldextran Hydrogel, mitt lere Dichte 1 * 1 ST
Catalog # MSPPSCBSBD200M
XAN TEC SPR Sensorchip. Biacore Series S kompatibel. Beschichtung: 200 nm Biotin derivatisiertes Carboxymethyldextran Hydrogel, mitt lere Dichte 1 * 1 ST
Catalog # MSPPSCBSBD200M
Frequently Bought Together