HEATER TOP LOADING COMPACT MODELS 2.9KW 1 * 1 items
Catalog # PRIOHTR/MID/C60
Anbieter: Priorclave
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Anbieter: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Anbieter: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
VWR® Serological Pipette, Standard Line
Anbieter: VWR Collection
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Anbieter: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals
VWR® Facial Tissues
Anbieter: VWR Collection
VWR®, Nitrile Gloves
Anbieter: VWR Collection
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
VWR®, Autoclavable Bags
Anbieter: VWR Collection