ContactUsLinkComponent
PCB for H2O2 decontamination. Also requires MCO-170UVS-PE and MCO-170EL-PE fitted plus MCO-50HP-PE and 1x MCO-50H2O2-PE 1 * 1 items
  SANYMCO-50HB-PW
PCB for H2O2 decontamination. Also requires MCO-170UVS-PE and MCO-170EL-PE fitted plus MCO-50HP-PE and 1x MCO-50H2O2-PE 1 * 1 items
  SANYMCO-50HB-PW
Frequently Bought Together