ContactUsLinkComponent
Module IQ/microcontroller for system 2020, for up to 20 IQ sensors, with MODBUS connection, automatic air pressure compensation as well as USB interface (IP 66) and Ethernet interface (IP 66) (Please order separately D-SUB connection ADA/D-SUB 902888 1 * 1 items
Catalog # WTWO471022
Supplier:  WTW ONLINE
Module IQ/microcontroller for system 2020, for up to 20 IQ sensors, with MODBUS connection, automatic air pressure compensation as well as USB interface (IP 66) and Ethernet interface (IP 66) (Please order separately D-SUB connection ADA/D-SUB 902888 1 * 1 items
Catalog # WTWO471022
Supplier:  WTW ONLINE
Frequently Bought Together