SureLight Glow - Firefly Luciferase Cell Viability Assay 1 * 100 Tests
Catalog # CAYM34972-100
Supplier: Cayman Chemical
SureLight Glow - Firefly Luciferase Cell Viability Assay 1 * 100 Tests
Catalog # CAYM34972-100
Supplier: Cayman Chemical
Frequently Bought Together
n-Hexane ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Benzoic acid methyl ester
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
n-Pentane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
1-Butanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Turpentine oil
Supplier: Roth Carl
Videojet Technologies, OPLOSMIDDEL 750 M
Supplier: MARKET SOURCE PART PROCESS
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals