ContactUsLinkComponent
SARS-CoV-2 Spike RBD His Tag (B.1.1.529/Omicron) (SPD-C522e) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - L ys 537 (Accession # QHD43416.1 (G339D S371L S373P S375F K417N N440K G446S S477N T478K E484A Q493K G496S Q4 05H)). The 1 * 100 µG
Catalog # ACRBSPDC522E-100UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike RBD His Tag (B.1.1.529/Omicron) (SPD-C522e) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - L ys 537 (Accession # QHD43416.1 (G339D S371L S373P S375F K417N N440K G446S S477N T478K E484A Q493K G496S Q4 05H)). The 1 * 100 µG
Catalog # ACRBSPDC522E-100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together