AABLOCKS 3ALPHA-HYDROXY-5BETA-CHOLAN 24-OIC ACID N-[CARBOXYMETHYL]AMIDE 3-SULFATE DISODIUM SALT 1 * 5 mg
Catalog # MSPPAA00EEJH-5MG
Supplier: MARKET SOURCE PART PROCESS
AABLOCKS 3ALPHA-HYDROXY-5BETA-CHOLAN 24-OIC ACID N-[CARBOXYMETHYL]AMIDE 3-SULFATE DISODIUM SALT 1 * 5 mg
Catalog # MSPPAA00EEJH-5MG
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals