5-OXASPIRO[2.5]OCTAN-8-ONE 1 * 1 g
Catalog # PHBLPBLJ8159-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
p-Xylene, EMSURE® ISO for analysis, Supelco®
Supplier: Merck
Acetonitrile, Prepsolv® for preparative chromatography
Supplier: Merck
Dimethyl sulphoxide, extra dry 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate 99.5% for analysis
Supplier: Thermo Fisher Scientific
Acetone 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
n-Heptane, anhydrous (max. 0.003% H₂O)
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99%
Supplier: Thermo Fisher Scientific
1,4-Dioxane, anhydrous (max. 0.003% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals