alpha-KETOISOVALERIC ACID Sodium salt 1 * 0,5 g
Catalog # CAMBCDLM-10647-0.5
Supplier: CAMBRIDGE ISOTOPE LABS
alpha-KETOISOVALERIC ACID Sodium salt 1 * 0,5 g
Catalog # CAMBCDLM-10647-0.5
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine 85% Ph. Eur.
Supplier: VWR Chemicals
Toluene ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Supplier: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99.5% ACS
Supplier: Thermo Fisher Scientific