CAMBRIDGE TECH 1-Axis scanning set, CRS12K galvo and CRS servo driver, 5mm OVAL mirror (450-7000nm coating, Be Butterfly substrat e), 10 degrees (optical PtP) field, 30cm cable, 12V power supply, +5V command input. 1 * 1 ST
Catalog # MSPP6SC12KA012-04Y
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together