Order Entry
Switzerland
Orders LinkContactUsLinkComponent
ISO warning labels | Avantor

ISO warning labels

ISO warning label - General Warning Symbol, Black/yellow, 19,05×19,05 mm, 250 labels/roll
Katalognr. GAINWL-009-0.25R
Anbieter:   GA
ISO warning labels
Katalognr. GAINWL-009-0.25R
Anbieter:  GA
ISO warning labels
ISO warning labels

ISO warning label - General Warning Symbol, Black/yellow, 19,05×19,05 mm, 250 labels/roll

Preis
SFr. 55.70
Einloggen für Verfügbarkeit
1 * 1 ROU
Product Details & Documents

Pre-printed general warning labels for the identification of equipment, containers, and material that may pose a safety risk in the laboratory environment.

  • Withstand high-temperature conditions up to 100° C (212° F)
  • Pre-printed with full color, yellow on silver background, design
  • Durable thermoplastic material is resistant to wear and tear, water, mild chemicals, and detergents
  • Resistant to gamma irradiation (tested to up to 50kGy)

Coated with a glove-friendly adhesive, these waterproof thermoplastic labels are perfect for labeling water baths, heat blocks, autoclave containers, and instruments or tools with moving parts.

Technische Daten
  • Colour:
    Black/yellow
  • Description:
    ISO warning label - General Warning Symbol
  • Dimensions:
    19,05×19,05 mm
  • Seq:
    03
  • Packed:
    250 labels/roll