Orders LinkContactUsLinkComponent
Volumetric flasks, class B
100 ml, 14/23, 0,20 ± ml
Catalog # VITL673941
Supplier:   VITLAB
Volumetric flasks, class B
Catalog # VITL673941
Supplier:  VITLAB
Volumetric flasks, class B
Volumetric flasks, class B
100 ml, 14/23, 0,20 ± ml
Price
SFr. 45.00
Log in for availability
1 * 2 ST
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
Specifications
  • NS:
    14/23
  • Capacity:
    100 ml
  • Seq:
    04
  • Tolerance:
    0,20 ± ml