Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
50 ml, 10/19, 0,12 ± ml
Catalog # VITL672941
Supplier:   VITLAB
Volumetric flasks, class B
Catalog # VITL672941
Supplier:  VITLAB
Volumetric flasks, class B
Volumetric flasks, class B
50 ml, 10/19, 0,12 ± ml
Price
SFr. 42.10
Log in for availability
1 * 2 ST
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
Specifications
  • NS:
    10/19
  • Capacity:
    50 ml
  • Seq:
    03
  • Tolerance:
    0,12 ± ml