Orders LinkContactUsLinkComponent
Digital microscopes, Emspira 3
Digital microscopes, Emspira 3
Emspira 3 digital microscope - stand-alone configuration incl. 4K bundle, 12 MP
Catalog # 630-3382
Supplier:   LEICA MICROSYSTEMS
Digital microscopes, Emspira 3
Catalog # 630-3382
Supplier:  LEICA MICROSYSTEMS
Digital microscopes, Emspira 3
Digital microscopes, Emspira 3
Emspira 3 digital microscope - stand-alone configuration incl. 4K bundle, 12 MP
Price
SFr. 12’727.97
Log in for availability
1 * 1 SET
Product Details & Documents
Emspira 3 digital microscope empowers users to streamline inspection processes, cover inspection needs flexibly, and work with confidence. An ideal partner for manipulation, quality control, inspection and research applications.
  • Flexibility thanks to the modular design and large variety of accessories
  • Ease of use - create overlays and reference images for comparison
  • Future proof - capture directly to USB stick or network folder or send images via email
  • Intuitive, customisable UI with mouse controls
  • High quality, robust and antimicrobial housing AgTreat™ - IP 21
Streamline your inspection process
Emspira 3 helps you to perform comprehensive visual inspection tasks with no need for a PC, giving you in addition the advantage of less clutter. Its integrated on-screen display (OSD) provides intuitive tools for stand-alone operation.
Directly compare to references with a single click
Pass / fail decision making is facilitated by comparing the live image directly to reference images or customized overlays with a single click. It is especially helpful for recurring tasks, e.g. checking if parts or components are within specified tolerances.
Keep your data easily available at all times
Minimize the risk of data loss by keeping results and data on your local network for worry-free storage. This capability gives you the confidence that all relevant data is available when you need it.
Adaptable for your different tasks
Customize Emspira 3 to the demands of many samples and easily adapt it to the changing needs of specific applications, like R and D and failure analysis, thanks to its modular design and large variety of accessories. Depending on your tasks, you choose how to store images - on USB drives or in network folders.
Just use it - designed for everyone
Emspira 3 is designed for users of any skill level. You can operate the system intuitively with minimal training, saving you time and effort. Work according to your preferred style - using a mouse, keyboard, touch display, and hand / foot switch.
Ergonomic workplace
Emspira 3 as a digital microscope without eyepieces offers ergonomic benefits. Users can inspect in a comfortable way with less strain when viewing fast live images (up to 60 fps) in 4K resolution on a monitor.
Ordering information: Please note that the power cord has to be ordered separately (except for the stand-alone configurations). Two-year warranty.
Delivery information: 630-3381 includes: Emspira 3 Microscope, Objective 1.0x Planapo Z-series, Adapter-Telecentric (0.8× and 1.0× Planapo), Adapter M60 body to M42 objective thread, Focus drive 300 mm M-Series Routine, Microscope carrier, Incident light base (Medium with AntiShock), Leica LED3000 RL (58 mm), Diffusor for LED3000 RL, Power Supply for LED3000/LED5000, Dust cover (40×35×60 cm, antistatic), 10"-touch HDMI monitor with standard column mount, Emspira 3 Standalone Kit, Power cord.
630-3382 includes: Emspira 3 - 4k bundle (incl. microscope, 4K monitor and accessories), Objective 1.0x Planapo Z-series, Adapter-Telecentric (0.8× and 1.0× Planapo), Adapter M60 body to M42 objective thread, Focus drive 300 mm M-Series Routine, Microscope carrier, Incident light base (Medium with AntiShock), Leica LED3000 RL (58 mm), Diffusor for LED3000 RL, Power Supply for LED3000/LED5000, Dust cover (40×35×60 cm, antistatic), Power cord.
Specifications
  • Resolution:
    12 MP
  • Description:
    Emspira 3 digital microscope - stand-alone configuration incl. 4K bundle
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Model
    Availability
    Price
  • Leica LED3000 SLI, spotlight illumination
    Leica LED3000 SLI, spotlight illumination
    LEIM10450508
    For digital microscope Emspira 3
    Log in for availability
    1 * 1 ST - SFr. 942.60