Filtration kits, Sartoclear® Dynamics Lab V
Sartoclear® Dynamics Lab V, 10 g, Asymmetric PES, 1000 ml, 0,22 µm
Katalognr. 514-1338
Anbieter: Sartorius
Filtration kits, Sartoclear® Dynamics Lab V
Katalognr. 514-1338
Anbieter: Sartorius
Direkt zu
Product Details & Documents
Sartoclear Dynamics® Lab V kits enable clarification and sterile filtration to be performed in a single step.
- Clarify high-density mammalian cell culture in minutes
- Out-of-the-box solution
- Fast and effortless
- No need for centrifugation
These kits simplify the process by fully eliminating the centrifugation step otherwise needed for clarification. As a result, they enable harvested cell cultures to be efficiently clarified and sterilised in minutes – quickly and easily.
Sartoclear Dynamics® Lab V kits are designed for simultaneous clarification and sterile filtration of ≻15 ml to 1 L of mammalian cell culture broth to prepare samples for subsequent downstream processing steps, such as protein purification and concentration. The convenient kits combine pouches of filter aid for clarification and vacuum filtration units for sterile filtration.
Technische Daten
- Membrane:Asymmetric PES
- Pore size:0,22 µm
- Capacity:1000 ml
- Description:Sartoclear® Dynamics Lab V, 10 g
- Seq:08
- Cat. No.:514-1338
- Environmental certification:Renewable Energy Certificate
- Environmentally Preferable:Low Manufacturing Impact



