Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 4×96 preps, 1 KIT
Catalog # OMEGM4030-01
Supplier:   OMEGA BIO-TEK
Mag-Bind® Universal Pathogen Core Kit
Catalog # OMEGM4030-01
Supplier:  OMEGA BIO-TEK
CAS Number:  
Mag-Bind® Universal Pathogen Core Kit

Universal pathogen kit, 4×96 preps, 1 KIT

CAS Number:  
Price
1 840.00 SFr.
Log in for availability
1 * 1 KIT
Product Details & Documents

Isolate host genomic, bacterial, fungal spore, and viral DNA and viral RNA from tissue, urine, serum, and fecal samples using magnetic beads.

  • Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications

The Mag-Bind® Universal Pathogen Core Kit allows rapid and reliable isolation of high-quality host genomic DNA, gram-positive and negative bacterial DNA, fungal spore DNA, viral DNA, and viral RNA from tissue, urine, serum, and fecal samples. The system incorporates a recommended enzymatic lysis which allows for fully automated extraction post-lysis via Hamilton STAR™, Thermo KingFisher™ Flex, Applied Biosystems® MagMAX™, Qiagen Biosprint, Tecan Fluent® Automation Workstation, Tecan Freedom EVO®, and other liquid handling instruments. There are no organic extractions, thus reducing plastic waste and hands-on time and multiple samples can be processed in parallel. The system combines the Mag-Bind® technology with RBB Buffer to eliminate PCR inhibiting compounds within the samples. Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Universal pathogen kit
  • Contents:
    4×96 preps
  • Cat. No.:
    OMEGM4030-01
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Magnetic beads
  • Sample type:
    Bacteria

Specifications