ContactUsLinkComponent
M6005: PCR Cycle Number Analysis MiniLab & Gel Electrophoresis Combo Reagent & Accessories for 10 Student Groups which includes: FastTaq PCR MasterMix (2X), Primer Set, Lambda DNA Sample, Sterile Nuclease-free Water, MiniOne DNA Marker, MiniOne 5X S 1 * 1 KIT
Katalognr. EMBIM6005TAE
Supplier:  EMBITEC
M6005: PCR Cycle Number Analysis MiniLab & Gel Electrophoresis Combo Reagent & Accessories for 10 Student Groups which includes: FastTaq PCR MasterMix (2X), Primer Set, Lambda DNA Sample, Sterile Nuclease-free Water, MiniOne DNA Marker, MiniOne 5X S 1 * 1 KIT
Katalognr. EMBIM6005TAE
Supplier:  EMBITEC
Frequently Bought Together