ContactUsLinkComponent
1% Agarose GelCups (TAE) w/o Stain: includes Ten GelCups with preweighed 1% Agarose in 0.5XTAEbuffer, 100 mL Tris-Acetate-EDTA (TAE) buffer concentrate 1 * 10 items
Catalog # EMBIM3251TAE
Supplier:  EMBITEC
1% Agarose GelCups (TAE) w/o Stain: includes Ten GelCups with preweighed 1% Agarose in 0.5XTAEbuffer, 100 mL Tris-Acetate-EDTA (TAE) buffer concentrate 1 * 10 items
Catalog # EMBIM3251TAE
Supplier:  EMBITEC
Frequently Bought Together